Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Solvents
▸
Isopropanols
▸
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
For nucleic acid precipitation | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Catalog Number
Quantity
GEEBP2618-212
2.5 L
Catalog number
GEEBP2618-212
Price (SGD)
150.00
Each
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (SGD)
150.00
Each
Add to cart
Specifications
Chemical Name or Material
Isopropanol
pH
7
Melting Point
-89.5°C
Absorbance
0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
CAS
67-63-0
View more
For nucleic acid precipitation
Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids.