Check Order Status
Ammonium Acetate, Optima™ LC/MS Grade, Fisher Chemical™
Element Search
Structure Search
Thermo Scientific Chemicals
Ammonium Acetate, Optima™ LC/MS Grade, Fisher Chemical™
Ammonium Acetate, Optima LC/MS Grade, Fisher Chemical, CAS: 631-61-8, White, Melting Point: 114 deg.C, Molecular Formula: C2H7NO2, pH: 7, Quantity: 50g, Formula Weight: 77.08g/mol, MDL Number: 13066, Physical Form: Solid
Have Questions?
Catalog number
GEEA114-50
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Melting Point
114°C
CAS
631-61-8
Color
White
pH
7
Physical Form
Solid
View more
Ammonium Acetate, Optima LC/MS Grade, Fisher Chemical, CAS: 631-61-8, White, Melting Point: 114 deg.C, Molecular Formula: C2H7NO2, pH: 7, Quantity: 50g, Formula Weight: 77.08g/mol, MDL Number: 13066, Physical Form: Solid