1x1 image pixel for data collection
Isotemp™ Shaking Water Baths
Isotemp™ Shaking Water Baths
Actual product may vary
Isotemp™ Shaking Water Baths
Isotemp™ Shaking Water Baths
Foamtec
Isotemp™ Shaking Water Baths
With a capacity up to 27 liters (7 gallons) shaking water baths have intuitive digital control and are easy-to-use.
Have Questions?
Catalog NumberModelDescriptionCapacity (Metric)Electrical Requirements
FTNFSSWB15Isotemp SWB 1515L Shaking Water Bath15 L100/115 V 200/230 V 50/60 Hz
Catalog number FTNFSSWB15
Price (SGD)
5,453.00
EA
Estimated availability date 23-Oct-2025
Add to cart
Model:
Isotemp SWB 15
Description:
15L Shaking Water Bath
Capacity (Metric):
15 L
Electrical Requirements:
100/115 V 200/230 V 50/60 Hz
Price (SGD)
5,453.00
EA
Add to cart
With a capacity up to 27 liters (7 gallons) Fisher Scientific™ Isotemp™ shaking water baths have intuitive digital control and are easy-to-use. These versatile shaking water baths can be used across a wide array of applications from life science and QA/QC. Count on the latest in technology, features and options to deliver the ideal combination of performance, ease of use, and reliability.
  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with temperature and shaking speed presets
  • New audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor and pump
  • Global voltage – each bath works anywhere in the world exactly the same 
Specifications
Chamber Material304 Stainless Steel
Controller TypeDigital
Description15L Shaking Water Bath
Capacity (Metric)15 L
Certifications/ComplianceUL, CE, FDA Class I
Temperature RangeAmbient +5° to 100°C
DisplayMonochrome LCD
Heating Capacity1200 w
IncludesStainless Steel Gable Cover, Shaking Tray
MaterialEpoxy Powder-coated Cold Rolled Steel
ModelIsotemp SWB 15
Temperature Presets3
Temperature Stability±0.1°C
Temperature Uniformity±0.05°C
Dimensions (L x W x H)15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm)
Electrical Requirements100/115 V 200/230 V 50/60 Hz
Amperage9.0-10.5 / 4.3-5.0 (120V/230V)
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link