Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Model |
|---|---|---|
| FTNFSGPD02 | 2 L | Isotemp GPD 02 |
| FKS15365877 | 2 L | Isotemp GPD 2S |
| FTNFSGPD05 | 5 L | Isotemp GPD 05 |
| FTNFSGPD10 | 10 L | Isotemp GPD 10 |
| FTNFSGPD15D | 15 L (5 L/10 L) | Isotemp GPD 15D |
| FTNFSGPD20 | 20 L | Isotemp GPD 20 |
| FTNFSGPD28 | 28 L | Isotemp GPD 28 |