1x1 image pixel for data collection
Isotemp™ R28 Refrigerated and Heated Bath Circulators: 6.8-8.6L, 230V/50Hz
Isotemp™ R28 Refrigerated and Heated Bath Circulators: 6.8-8.6L, 230V/50Hz
Actual product may vary
Isotemp™ R28 Refrigerated and Heated Bath Circulators: 6.8-8.6L, 230V/50Hz
For laboratory and industrial applications.
Have Questions?
Catalog NumberTemperature RangeModelPump Type
FTN211333100-28°C to +200°CIsotemp 6200 R28Force and Suction
Catalog number FTN211333100
Price (SGD)
16,614.00
EA
Estimated availability date 23-Oct-2025
Add to cart
Temperature Range:
-28°C to +200°C
Model:
Isotemp 6200 R28
Pump Type:
Force and Suction
Price (SGD)
16,614.00
EA
Add to cart
Low-profile model enables easy bath access when lab counter height presents a challenge. Seamless stainless-steel tanks guarantee durability and easy cleaning. Robust refrigeration system ensures that samples and applications can be cooled quickly. Fine-tuned refrigeration metering allows for excellent temperature stability at all times.
  • All bath drains are located on the front of the unit with an integrated valve
Green Tip: For less demanding applications, power consumption can be lowered by utilizing the energy savings mode
Specifications
Pump TypeForce and Suction
Temperature Range-28°C to +200°C
ModelIsotemp 6200 R28
Voltage230 V
Frequency50 Hz
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link