Isotemp™ Digital-Control Water Baths: Model 215
Product Image
Isotemp™ Digital-Control Water Baths: Model 215
Ideal for industrial, clinical, pharmaceutical or biomedical procedures
Have Questions?
Catalog number FTN1546216Q
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Ideal for industrial, clinical, pharmaceutical or biomedical applications
Microprocessor PID Control
  • ±0.24°C uniformity, ±0.5°C accuracy and stability at 37°C
  • Display shows °C or °F; easy to read from across the room
Safety Features
  • Low profile and front-panel controls for easier access, safer operation
  • Dual thermostats guard against thermal runaway
  • Hinged acrylic or stainless-steel gable covers have fins that keep hands away from hot vapors
  • Covers stay open at 90° or, with quick-release hinge, lift off completely for use with large glassware
  • Heating elements will not burn out if bath accidentally runs dry
Construction
  • Corrosion-resistant exterior with tough, easy-to-clean acrylic enamel coating
  • Temperature-resistant plastic rim insulates, resists leaks, keeps its shape and won't dry out or crack like caulked seals
  • Exterior stays cool even after hours of operation Deep-drawn, stainless-steel reservoirs and perforated sample trays prevent leaks, guard against rust and contamination
  • Rounded reservoir corners and sample trays with finger holes allow for easy removal, make cleaning easier
  • Choice of acrylic or stainless-steel covers
Model 215
  • With drain pump to empty reservoir
Specifications
Controller TypeMicroprocessor PID
Capacity (Metric)5 and 10 L
Certifications/ComplianceCSA
Temperature RangeAmbient to 100°C
For Use With (Application)Industrial, clinical, pharmaceutical or biomedical procedures
IncludesAcrylic cover
Model215
Temperature Accuracy±0.5°C
Temperature Resolution±0.1°C
Temperature Stability±0.5°C
Warranty1 Year
Dimensions (L x W x H)16.53 x 24.38 x 8.88 in. (42 x 62 x 23 cm)
Length (English)16.53 in.
Length (Metric)42 cm
Width (English)24.38 in.
Width (Metric)62 cm
Height (English)8.88 in.
Height (Metric)23 cm
Shipping Weight (English)24.5 lb.
Shipping Weight (Metric)11.1 kg
Voltage230 V
Amperage8.3 A
Frequency60 Hz
Unit SizeEA