▸
pH and Electrochemistry
▸
pH and Electrochemistry Meter Accessories
▸
Multiparameter Meter Accessories
▸
RS232 Thermal Printer for accumet™ AB and XL Series Benchtop Meters
Actual product may vary
RS232 Thermal Printer for accumet™ AB and XL Series Benchtop Meters
Convenient and compact
Have Questions?
Catalog Number
ForUseWithEquipment
FSP13-637-690
Catalog number
FSP13-637-690
Price (SGD)
1,702.00
Each
Add to cart
Price (SGD)
1,702.00
Each
Add to cart
Small Footprint Thermal Printer
Compact 5.25 in. L x 3.75 in. W
Global power supply for 100/240VAC
Operating Temperature: 0° to 50°C (41° to 104° F)
Unit Weight: 0.75 lb. (0.34kg)
Serial RS-232
Paper Width: 2.3 in. (58.4mm)
Print Width: 1.9 in. (48.2mm) @ 384 dots per line
Printer cable (13-637-681) and paper rolls (13-637-691) available separately
Specifications
Dimensions (L x W)
5.25 x 3.75 in. (0.13m x 0.09 m)
Length (English)
5.25 in.
Length (Metric)
0.13 m
Voltage
100/240 V
Width (English)
3.75 in.
Width (Metric)
0.09 m