▸
pH and Electrochemistry
▸
pH and Electrochemistry Meter Accessories
▸
Multiparameter Meter Accessories
▸
RS232 Thermal Printer Cable for accumet™ AB and XL Series Benchtop Meters
Actual product may vary
RS232 Thermal Printer Cable for accumet™ AB and XL Series Benchtop Meters
RS-232 compaible
Have Questions?
Catalog Number
For Use With (Equipment)
FSP13-637-681
Accumet AB, XL Series Meters
Catalog number
FSP13-637-681
Price (SGD)
-
For Use With (Equipment):
Accumet AB, XL Series Meters
RS232 Thermal printer for Accumet™ AB and XL series bench-top meter
RS232 thermal printer cable
Use with: AB and XL Series
Specifications
Type
Printer Cable
Includes
RS-232 thermal printer cable
For Use With (Equipment)
Accumet AB, XL Series Meters