Check Order Status
Eutech Handheld Meter Calibration Solution, 480 mL 111.8 mS/cm KCl
Element Search
Structure Search
Eutech Handheld Meter Calibration Solution, 480 mL 111.8 mS/cm KCl
Periodic calibration with accurate standards is recommended for meter
Have Questions?
Catalog Number
Quantity
FSP01X211244
Catalog number
FSP01X211244
Price (SGD)
-
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7
Specifications
Chemical Name or Material
Inorganic Salts
CAS
7447-40-7
Recommended Storage
Keep container tightly closed. Keep in cool dark area away from light
Odor
Odorless
Density
1.0 g/cm³ at 25°C
View more
Periodic calibration with accurate standards is recommended for meter
480 mL 111.8 mS/cm KC calibration solution
Solutions packed in 480mL durable plastic bottles
Economical and convenient for laboratory use.