Check Order Status
Eutech Handheld Meters Calibration Solution, 1413μS/cm KCl
Element Search
Structure Search
Eutech Handheld Meters Calibration Solution, 1413μS/cm KCl
Periodic calibration with accurate standards is recommended for meters
Have Questions?
Catalog number
FSP01X211207
Price (SGD)
-
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7
Specifications
Chemical Name or Material
Inorganic Salts
Name Note
Nil
Packaging
1 case
CAS
7447-40-7
Recommended Storage
Keep container tightly closed. Keep in cool, dark area away from light.
View more
1413μS/cm KCl calibration solution
Solutions packed in 480mL durable plastic bottles
Economical and convenient for laboratory use.