1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT
1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT
1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT
1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT
Thermo Scientific Chemicals
1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT
CAS: 108-65-6 | C6H12O3 | 132.159 g/mol
Have Questions?
Catalog NumberQuantity
FSKL15459.AU1000 mL
Catalog number FSKL15459.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or Material1,2-Propanediol monomethyl ether acetate
Name Notestabilized with 50ppm BHT
CAS108-65-6
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapor.
View more
1,2-Propanediol monomethyl ether acetate is used as a solvent for paints, inks, lacquers, varnishes, cleaners and coatings. It is also used as a solvent for degreasing circuit boards and in food contact applications. Further, it is used in photoresist formulations in the semiconductor industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Propanediol monomethyl ether acetate is used as a solvent for paints, inks, lacquers, varnishes, cleaners and coatings. It is also used as a solvent for degreasing circuit boards and in food contact applications. Further, it is used in photoresist formulations in the semiconductor industry.
Solubility
Miscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, reducing agents, acids and alkalis.
RUO – Research Use Only
General References:
  1. Baysal, G.; Önder, S.; Göcek, İ.; Trabzon, L.; Kızıl, H.; Kök, F. N.; Kayaoğlu, B. K. Design and fabrication of a new nonwoven-textile based platform for biosensor construction. Sens. Actuators, B 2015, 208, 475-484.
  2. Charlton, M. R.; Dylla, A. G.; Stevenson, K. J. Direct Evidence of a Chemical Conversion Mechanism of Atomic-Layer-Deposited TiO2 Anodes During Lithiation Using LiPF6 Salt. J. Phys. Chem. 2015, 119 (51), 28285-28291.