Water, Endotoxin-free
Water, Endotoxin-free
Water, Endotoxin-free
Water, Endotoxin-free
Thermo Scientific Chemicals
Water, Endotoxin-free
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Catalog NumberQuantity
FSKJ65589.K21 L
Catalog number FSKJ65589.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialWater, Endotoxin-free
pH7
CAS7732-18-5
Melting Point32°F
Boiling Point100°C
View more
Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.
Solubility
Miscible with methanol, acetone and ethanol.
RUO – Research Use Only
General References:
  1. Narayanan, D.; Geena, M. G.; Lakshmi, H.; Koyakutty, M.; Nair, S.; Menon, D. Poly-(ethylene glycol) modified gelatin nanoparticles for sustained delivery of the anti-inflammatory drug Ibuprofen-Sodium: An in vitro and in vivo analysis. Medicine 2013, 9 (6), 818-828.
  2. Madhubanti, B.; Kumar, M. N.; Mrinal, S. Toll-like receptor (TLR) 4 in mrigal (Cirrhinus mrigala) : Response to lipopolysaccharide treatment and Aeromonas hydrophila infection. Int. Res. J. Biological Sci. 2013, 2 (4), 20-27.