Diethylaminoethyl dextran
Diethylaminoethyl dextran
Diethylaminoethyl dextran
Diethylaminoethyl dextran
Thermo Scientific Chemicals
Diethylaminoethyl dextran
CAS: 9015-73-0
Have Questions?
Catalog NumberQuantity
FSKJ63781.1425 g
Catalog number FSKJ63781.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylaminoethyl dextran
CAS9015-73-0
ColorWhite
Recommended StorageAmbient temperatures
RTECS NumberHH9250000
View more
DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. The use of DEAE-Dextran as an efficient adjuvant in vaccine production is well established. For instance, DEAE-Dextran has been used for immunization of guinea pigs and swine with inactivated foot-and-mouth disease and rhesus monkeys with inactivated Venezulean equine encephalomyelitis virus, IVEE. The mechanism of action of DEAE-Dextran is not clear. The effect on the humoral response of rhesus monkeys to IVEE vaccine results in a typical IgM-IgG response. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. James H. McCutchan,; Joseph S. Pagano. Enhancement of the Infectivity of Simian Virus 40 Deoxyribonucleic Acid With Diethylaminoethyl-Dextran2. J Natl Cancer Inst. 1968, 41 (2),351-357..
  2. H Duc-Nguyen. Enhancing effect of diethylaminoethyl-dextran on the focus-forming titer of a murine sarcoma virus (Harvey strain). J Virol. 1968, 2 (6), 643-644.