Pectinase, Aspergillus niger
Pectinase, Aspergillus niger
Thermo Scientific Chemicals
Pectinase, Aspergillus niger
Used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation.
Have Questions?
Catalog NumberQuantity
FSKJ63408.MD250 mg
Catalog number FSKJ63408.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Recommended StorageAmbient temperatures
EINECS Number232-885-6
Quantity250 mg
FormPowder
Unit SizeEA
Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.