BIS-TRIS, 0.2M buffer soln., pH 6.0
BIS-TRIS, 0.2M buffer soln., pH 6.0
BIS-TRIS, 0.2M buffer soln., pH 6.0
BIS-TRIS, 0.2M buffer soln., pH 6.0
Thermo Scientific Chemicals
BIS-TRIS, 0.2M buffer soln., pH 6.0
CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol
Have Questions?
Catalog NumberQuantity
FSKJ62686.AK250 mL
Catalog number FSKJ62686.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBIS-TRIS
CAS6976-37-0
ColorColorless
Storage Note 12° to 8°C
Recommended StorageKeep cold