Hydroquinone bis(2-hydroxyethyl) ether, 95%
Hydroquinone bis(2-hydroxyethyl) ether, 95%
Hydroquinone bis(2-hydroxyethyl) ether, 95%
Hydroquinone bis(2-hydroxyethyl) ether, 95%
Thermo Scientific Chemicals
Hydroquinone bis(2-hydroxyethyl) ether, 95%
CAS: 104-38-1 | C10H14O4 | 198.218 g/mol
Have Questions?
Catalog NumberQuantity
FSKB24037.36500 g
Catalog number FSKB24037.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHydroquinone bis(2-hydroxyethyl) ether
CAS104-38-1
Melting Point100°C to 102°C
Recommended StorageAmbient temperatures
EINECS Number203-197-3
View more
Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. Protect from humidity and water. It is hygroscopic in nature. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. S. Kiran.; Roy Joseph. Synthesis and characterization of a noncytotoxic, X-ray opaque polyurethane containing iodinated hydroquinone bis(2-hydroxyethyl) ether as chain extender for biomedical applications. Journal of Biomedical Materials Research Part A. 2014, 102 (9), 3207-3215.
  2. Mei-Fei Wang.; Jun-Qin Qiao.; Zheng-Chun Qi.; Yu-Na Wei.;Hong-Zhen Lian.; Tao Ding.; Chong-Yu Shen. Separation and Identification of Relative Substances in Polyurethane Chain Extender Hydroquinone Bis(2-hydroxyethyl) Ether by High Performance Liquid Chromatography with Photodiode Array Detection and Electrospray Ionization Mass Spectrometry. Ind. Eng. Chem. Res. 2009, 48 (5), 2487-2492.