Hydrogen chloride, 4M in 1,4-dioxane, 99%
Hydrogen chloride, 4M in 1,4-dioxane, 99%
Thermo Scientific Chemicals
Hydrogen chloride, 4M in 1,4-dioxane, 99%
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
FSKA16935.JV2 Mol
Catalog number FSKA16935.JV
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 Mol
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrogen chloride
CAS7647-01-0
Health Hazard 1H225-H314-H331-H335-H350
Health Hazard 2GHS H Statement
H225-H331-H314-H351-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Health Hazard 3P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Hydrogen chloride, 4M in 1,4-dioxane is used in the preparation of N-methyl-O-benzoylhydroxylamine hydrochloride via reaction with benzoic acid, carbonyldiimidazole and N-methylhydroxylamine hydrochloride.