1,10-Phenanthroline monohydrochloride monohydrate, 99%
1,10-Phenanthroline monohydrochloride monohydrate, 99%
1,10-Phenanthroline monohydrochloride monohydrate, 99%
1,10-Phenanthroline monohydrochloride monohydrate, 99%
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrochloride monohydrate, 99%
CAS: 18851-33-7 | C12H11ClN2O | 234.683 g/mol
Have Questions?
Catalog NumberQuantity
FSKA16254.065 g
Catalog number FSKA16254.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1,10-Phenanthroline monohydrochloride monohydrate
CAS18851-33-7
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a tightly closed container in a cool, well-ventilated area away from air and moisture.
RUO – Research Use Only
General References:
  1. Liu, Y. L.; Xu, X. Y.; Sun, P. C.; Chen, T. H. N-doped porous carbon nanosheets with embedded iron carbide nanoparticles for oxygen reduction reaction in acidic media. Int. J. Hydrogen Energy 2015, 40 (13), 4531-4539.
  2. Vyprachticky, D.; Kankova, D.; Pokorna, V.; Kminek, I.; Dzhabarov, V.; Cimrova, V. Novel and Simple Synthesis of Brominated 1,10-Phenanthrolines. Aust. J. Chem. 2014, 67 (6), 915-921.