Sodium dodecatungstophosphate hydrate
Sodium dodecatungstophosphate hydrate
Sodium dodecatungstophosphate hydrate
Sodium dodecatungstophosphate hydrate
Thermo Scientific Chemicals
Sodium dodecatungstophosphate hydrate
CAS: 51312-42-6 | Na3O40PW12 | 2946.12 g/mol
Have Questions?
Catalog NumberQuantity
FSKA11575.1425 g
Catalog number FSKA11575.14
Price (SGD)
153.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
153.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium dodecatungstophosphate hydrate
Melting Point350°C
CAS51312-42-6
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335