1x1 image pixel for data collection
Dopamine hydrochloride, 99%
Dopamine hydrochloride, 99%
Dopamine hydrochloride, 99%
Dopamine hydrochloride, 99%
Thermo Scientific Chemicals
Dopamine hydrochloride, 99%
CAS: 62-31-7 | C8H12ClNO2 | 189.64 g/mol
Have Questions?
Catalog NumberQuantity
FSKA11136.065 g
Catalog number FSKA11136.06
Price (SGD)
101.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
101.00
EA
Add to cart
Specifications
Chemical Name or MaterialDopamine hydrochloride
CAS62-31-7
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
View more
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
Solubility
Soluble in water, methanol and dimethyl sulfoxide. Insoluble in ether, petroleum ether, chloroform, benzene and toluene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhai, C.; Li, D.; Li, L.; Sun, F.; Ma, H.; Liu, X. Experimental and theoretical study on the hydrogen bonding between dopamine hydrochloride and N,N-dimethyl formamide. Spectrochim. Acta, Part A. 2015, 145 (7), 500-504.
  2. Gosliga, J. M.; Barter, L. S. Cardiovascular effects of dopamine hydrochloride and phenylephrine hydrochloride in healthy isoflurane-anesthetized New Zealand White rabbits (Oryctolagus cuniculus). Am J Vet Res. 2015, 76 (2), 116-121.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link