1,2,4-Trichlorobenzene, 99%
1,2,4-Trichlorobenzene, 99%
1,2,4-Trichlorobenzene, 99%
1,2,4-Trichlorobenzene, 99%
Thermo Scientific Chemicals
1,2,4-Trichlorobenzene, 99%
CAS: 120-82-1 | C6H3Cl3 | 181.44 g/mol
Have Questions?
Catalog NumberQuantity
FSKA10130.0E2500 g
Catalog number FSKA10130.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Specifications
Chemical Name or Material1,2,4-Trichlorobenzene
Melting Point17°C
CAS120-82-1
Health Hazard 1H302-H315-H319
Health Hazard 2GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.
View more
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
Solubility
Miscible with ether, benzene, CS{2}.Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Bujak, M.; Podsiadło, M.; Katrusiak, A. Relations between compression and thermal contraction in 1,2,4-trichlorobenzene and melting of trichlorobenzene isomers. CrystEngComm 2015, 17 (18), 3446-3451.
  2. Dong, W. H.; Zhang, P.; Lin, X. Y.; Zhang, Y.; Tabouré, A. Natural attenuation of 1,2,4-trichlorobenzene in shallow aquifer at the Luhuagang's landfill site, Kaifeng, China. Sci. Total Environ. 2015, 505, 216-222.
  3. Su, G.; Huang, L.; Liu, S.; Lu, H.; Yang, F.; Zheng, M. The combined disposal of 1,2 4-trichlorobenzene and nitrogen oxides using the synthesized Ce0.2TiAlαOx microanomaterial. Catal. Sci. Technol. 2015, 5 (2), 1041-1051.