Di-n-butyltin dilaurate, 95%
Di-n-butyltin dilaurate, 95%
Di-n-butyltin dilaurate, 95%
Di-n-butyltin dilaurate, 95%
Thermo Scientific Chemicals
Di-n-butyltin dilaurate, 95%
CAS: 77-58-7 | C32H64O4Sn
Have Questions?
Catalog NumberQuantity
FSK071130.1425 g
Catalog number FSK071130.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-n-butyltin dilaurate
CAS77-58-7
Health Hazard 1H314-H317-H335-H336-H341-H360FD-H370-H372
Health Hazard 2GHS H Statement
H301-H331-H360-H341-H314-H318-H317
Toxic if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
View more
Di-n-butyltin dilaurate is used as a catalyst for the production of polyurethanes as well as for the transesterification reactions. It is involved in the vulcanization of silicones and a stabilizer in polyvinyl chloride (PVC). It acts as a rust inhibitor for polyurethanes, polyols, silicones and as a fuel additive.