Paraformaldehyde, 32% w/v aq. soln., methanol free
Paraformaldehyde, 32% w/v aq. soln., methanol free
Paraformaldehyde, 32% w/v aq. soln., methanol free
Paraformaldehyde, 32% w/v aq. soln., methanol free
Thermo Scientific Chemicals
Paraformaldehyde, 32% w/v aq. soln., methanol free
Solution is micro-filtered and supplied in ampules. | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Catalog NumberQuantity
FSK047377.9L2 x 10 mL
Catalog number FSK047377.9L
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 x 10 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialParaformaldehyde
CAS50-00-0
Health Hazard 1H302+H332-H311-H314-H317-H335-H341-H350
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
Storage Note 1Ampouled under argon
View more
Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.
RUO – Research Use Only