Hydrochloric acid, 6.0N Standardized Solution
Hydrochloric acid, 6.0N Standardized Solution
Hydrochloric acid, 6.0N Standardized Solution
Hydrochloric acid, 6.0N Standardized Solution
Thermo Scientific Chemicals
Hydrochloric acid, 6.0N Standardized Solution
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
FSK044921.K21 L
Catalog number FSK044921.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Health Hazard 1H290-H314-H335
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
CAS7732-18-5
View more
6.0N HClsolution acts as a standard solution in volumetric titrations. In addition, it has industrial applications such as chemical transformations in hydrochlorination of rubber and production of vinyl and alkyl halide. In the semiconductor industry, it is used to etch semiconductor crystal and to purify silicon.