Potassium chloride, 3M aq. soln.
Potassium chloride, 3M aq. soln.
Potassium chloride, 3M aq. soln.
Potassium chloride, 3M aq. soln.
Thermo Scientific Chemicals
Potassium chloride, 3M aq. soln.
For Refilling Calomel Electrodes | CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Catalog NumberQuantity
FSK043398.AK250 mL
Catalog number FSK043398.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium chloride
CAS7447-40-7
ColorColorless
CAS7732-18-5
Recommended StorageAmbient temperatures
View more
For refilling calomel electrodesPotassium chloride, 3M aq. solution is used for volumetric analysis in laboratories.