Calcium sulfate, anhydrous, 99%
Calcium sulfate, anhydrous, 99%
Calcium sulfate, anhydrous, 99%
Calcium sulfate, anhydrous, 99%
Thermo Scientific Chemicals
Calcium sulfate, anhydrous, 99%
CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Catalog NumberQuantity
FSK040144.30250 g
Catalog number FSK040144.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium sulfate, anhydrous
Melting Point1450°C
CAS7778-18-9
ColorWhite
Health Hazard 1H315-H319 : Causes skin irritation. Causes serious eye irritation.
View more
Used as Portland-cement retarder; tile and plaster; It is the source of sulfur and sulfuric acid; Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.