Diethyl ether, HPLC Grade, 99%, stab. with ethanol
Diethyl ether, HPLC Grade, 99%, stab. with ethanol
Actual product may vary
Diethyl ether, HPLC Grade, 99%, stab. with ethanol
Diethyl ether, HPLC Grade, 99%, stab. with ethanol
Thermo Scientific Chemicals
Diethyl ether, HPLC Grade, 99%, stab. with ethanol
Widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents. | CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Catalog NumberQuantity
FSK038990.M14 x 1 L
Catalog number FSK038990.M1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 x 1 L
It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Specifications
Chemical Name or MaterialDiethyl ether
Melting Point°C to 116°C
CAS60-29-7
Health Hazard 1H224-H302-H305-H335-H336-H373
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c
Recommended StorageAmbient temperatures
DOT InformationHazard Class: 3; Packaging Group: I
Density0.713 g/mL
RTECS NumberKI5775000
Boiling Point34.6°C
EINECS Number200-467-2
Quantity4 x 1 L
TSCAYes
Flash Point-45°C (-49°F)
Physical FormLiquid
Assay Percent Range99%
Molecular FormulaC4H10O
MDL NumberMFCD00011646
Synonymdiethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
Solubility InformationImmiscible with water,miscible with alcohol.
InChI KeyRTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILESCCOCC
IUPAC Nameethoxyethane
Molecular Weight (g/mol)74.12
PubChem CID3283
ChEBICHEBI:35702
Formula Weight74.12
GradeHPLC
Unit SizePK