Iodine, 0.1N Standardized Solution
Iodine, 0.1N Standardized Solution
Iodine, 0.1N Standardized Solution
Iodine, 0.1N Standardized Solution
Thermo Scientific Chemicals
Iodine, 0.1N Standardized Solution
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Catalog NumberQuantity
FSK035634.K21 L
Catalog number FSK035634.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialIodine
CAS7553-56-2
Health Hazard 1H335-H373
Health Hazard 3P260-P314-P501c
Recommended StorageAmbient temperatures
View more
Iodine, 0.1N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.