Rubidium acetate, 99.8% (metals basis)
Rubidium acetate, 99.8% (metals basis)
Rubidium acetate, 99.8% (metals basis)
Rubidium acetate, 99.8% (metals basis)
Thermo Scientific Chemicals
Rubidium acetate, 99.8% (metals basis)
CAS: 563-67-7 | C2H3O2Rb | 144.51 g/mol
Have Questions?
Catalog NumberQuantity
FSK012890.0910 g
Catalog number FSK012890.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRubidium acetate
CAS563-67-7
Melting Point246°C
Recommended StorageAmbient temperatures
EINECS Number209-255-4
View more
Rubidium acetate has been reported to modify the acid/base character of the catalyst supports like silica, alumina, and titania. It can be used in the preparation of nanosized rubidium ferrite, and as a promoter on the palladium-catalyzed synthesis of vinyl acetate in heterogeneous gas reaction. It is used in medical and in research laboratories.