Model 505 Sonic Dismembrator
Model 505 Sonic Dismembrator
Actual product may vary
Model 505 Sonic Dismembrator
High power, programmable system ideal for nanotechnology, cell disruption and homogenization applications.
Have Questions?
Catalog NumberIncludesVoltage
FQSFB505-220Generator, converter, 1/2 in. dia. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included220/230 V
Catalog number FQSFB505-220
Price (SGD)
-
Includes:
Generator, converter, 1/2 in. dia. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
Voltage:
220/230 V
The Fisher Scientific™ Model 505 Sonic Dismembrator offers the same programmable features as Model 120 but delivers substantially more power. High power, programmable system ideal for nanotechnology, cell disruption and homogenization applications.
  • Processes 0.2 to 1000mL
  • 500 watts
  • A variety of accessories are available including probes for individual samples and a Cup Horn for indirect sonication
  • Programmable from 1 second to 10 hours
  • Pulse mode (1 second to 59 seconds)
  • Energy display (watts and Joules)
  • Applications Include: Nanoparticle dispersion, mixing and homogenization
Specifications
Voltage220/230 V
Electrical Requirements220/230 V, 50/60 Hz
Hertz20 kHz (Ultrasonic Frequency)
Wattage500 W
Volume Range (Metric)0.5 to 500 mL
Dimensions (L x W x H)15 x 18 x 9 in.
Height (English)9 in.
Length (English)15 in.
Width (English)18 in.
For Use With (Application)Ideal for nano particle dispersion, mixing and homogenization
IncludesGenerator, converter, 1/2 in. dia. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
ModelFB505
Timer1 sec. to 10 hrs.
Unit SizeEA