CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
CO<sub>2</sub> and LN<sub>2</sub> Backup Systems for Ultra-Low Temperature Freezers
Actual product may vary
Thermo Scientific
CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
FLN4567LEThermo Scientific TSX SeriesLN2 Backup, Field Installed
1960446TSE Series, Forma 7000 Series, HFU B Series, HFC Series, Revco CxF Series, TSC SeriesCO2 Backup, Field Installed
FLN4567
also known as AVL
Revco UxF, HERAfreeze HFU-T, Thermo Scientific TSU SeriesLN2 Backup, Field Installed
FCO4567LEThermo Scientific TSX SeriesCO2 Backup, Field Installed
Catalog number FLN4567LE
Price (SGD)
5,906.00
EA
Add to cart
For Use With (Equipment):
Thermo Scientific TSX Series
Type:
LN2 Backup, Field Installed
Price (SGD)
5,906.00
EA
Add to cart
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:
  • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
  • CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.
Specifications
TypeLN2 Backup, Field Installed
Temperature Range-80°C
For Use With (Equipment)Thermo Scientific TSX Series
HoldsBoxes (300)
Unit SizeEA