Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Actual product may vary
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Use Thermo Scientific™ Manual Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Have Questions?
Catalog NumberFor Use With (Equipment)Type
FKS17374163
also known as FKS17374163  
13 mm Crimp VialsManual Decrimper
Catalog number FKS17374163
also known as FKS17374163  
Price (SGD)
1,183.00
EA
Add to cart
For Use With (Equipment):
13 mm Crimp Vials
Type:
Manual Decrimper
Price (SGD)
1,183.00
EA
Add to cart
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
Specifications
TypeManual Decrimper
For Use With (Equipment)13 mm Crimp Vials
MaterialStainless Steel/Aluminum
Product LineSureSTART Level 2
Unit SizeEA