Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Connection Type |
---|---|---|
FKS17171936 | 1 mL | Luer Lock |
FIS14-955-456 | 1 mL | Luer Lock |
FKS15849152 | 1 mL | Luer Lock |
FIS14-955-464 | 1 mL | Luer Lock |
FIS14-955-457 | 3 mL | Luer Lock |
FKS15859152 | 3 mL | Luer Lock |
FIS14-955-458 | 5 mL | Luer Lock |
FKS15869152 | 5 mL | Luer Lock |
FIS14-955-459 | 10 mL | Luer Lock |
FIS14-955-460 | 20 mL | Luer Lock |
FKS15889152 | 20 mL | Luer Lock |
FIS14-955-461 | 60 mL | Luer Lock |
FKS15899152 | 60 mL | Luer Lock |
FIS14-955-450 | 1 mL | Leur Slip |
FIS14-955-462 | 1 mL | Leur Slip |
FIS14-955-454 | 20 mL | Leur Slip |
FKS15829152 | 20 mL | Leur Slip |
FKS15839152 | 60 mL | Leur Slip |