Microplate Centrifuge
Microplate Centrifuge
Actual product may vary
Microplate Centrifuge
For fast spindowns of PCR microplates. The Fisherbrand Microplate Centrifuge has a small footprint with an easy-access removable rotor for easier cleaning.
Have Questions?
Catalog NumberCapacity
FKS163113382 Standard PCR Microplates
Catalog number FKS16311338
Price (SGD)
4,304.00
EA
Add to cart
Capacity:
2 Standard PCR Microplates
Price (SGD)
4,304.00
EA
Add to cart
  • Accommodates two standard-sized PCR microplates
  • Easily spins down droplets and condensation
  • Easy insertion and removal of plates
  • Swing-out rotor design eliminates need for plate seals
  • Digital display
  • 2500 rpm/850 x g
  • Electronic brake
Specifications
Capacity2 Standard PCR Microplates
Max. Speed2,500 rpm
Max. RCF850 x g
DisplayDigital
Height (Metric) Exterior140 mm
Width (Metric) Exterior289 mm
Certifications/ComplianceCE marked
IncludesUnit and swing-out rotor, EU/UK power cord, instruction manual
Depth (Metric)356 mm
Rotor IncludedYes
RefrigeratedNo
Run Time1 sec. to 10 min.
Unit SizeEA