Polypropylene Clear Autoclave Bags
Actual product may vary
Fisherbrand
Polypropylene Clear Autoclave Bags
Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Model
FKS16297961410 x 630 mmBiohazard Waste Identification
FKS16247971310 x 660 mmBiohazard Waste Identification
FKS16207971620 x 780 mmBiohazard Waste Identification
FKS16237971620 x 780 mmPlain without Print
Catalog number FKS16297961
Price (SGD)
-
Dimensions (L x W):
410 x 630 mm
Model:
Biohazard Waste Identification
Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.
  • For high temperature sterilisation (134°C) to a maximum of 140°C
  • 40μm thickness
  • High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
  • For safe disposal of biohazard waste
  • Specifically designed for contaminated waste disposal in autoclaves or incinerators
  • Printed with a biohazard warning
Specifications
ColorClear
Dimensions (L x W)410 x 630 mm
ModelBiohazard Waste Identification
Quantity100/Pk.
Width (Metric)410 mm
AutoclavableAutoclavable
IncludesBiohazard Warning
Thickness40 μm
MaterialPolypropylene
Temperature (Metric)134°C to 140°C
Unit SizePack