Unisex White Stud Front Lab Coat
Unisex White Stud Front Lab Coat
Actual product may vary
Fisherbrand
Unisex White Stud Front Lab Coat
Machine-washable, polyester/cotton, unisex asymmetrical stud-front laboratory coats feature stretch cuffs for comfort and three useful pockets.
Have Questions?
Catalog NumberSize
FKS15851134Medium
Catalog number FKS15851134
Price (SGD)
102.00
EA
Add to cart
Size:
Medium
Price (SGD)
102.00
EA
Add to cart
  • Unisex asymmetrical stud front laboratory coat
  • Stretch cuffs for comfort
  • Two hip pockets, one chest pocket
  • Machine washable and industrially launderable
  • Unisex
  • Length 43ins/110cm
  • 245gsm 67% polyester/33% cotton
  • Available in a choice of colours in the wider product portfolio
Specifications
Product TypeGeneral Purpose Lab Coat
SizeMedium
GenderUnisex
MaterialPolycotton
ColorWhite
Closure TypePress Studs
Cuff StyleKnit
DisposableReusable
SterilityNon-sterile
No. of Pockets3
Collar TypeMandarin
Size (English) Chest40 in.
IncludesAssymetric stud front, two hip and one chest pockets
Length (English)43 inch
Length (Metric)110 cm
Unit SizeEA