Bottle Carrying System for DURAN™ GL-45 Laboratory Bottles
Bottle Carrying System for DURAN™ GL-45 Laboratory Bottles
Actual product may vary
Bottle Carrying System for DURAN™ GL-45 Laboratory Bottles
For easy and safe carrying and pouring of GL 45 laboratory glass bottles.
Have Questions?
Catalog NumberHoldsColor
FKS15826931Bottle (5 L)Yellow
Catalog number FKS15826931
Price (SGD)
489.00
EA
Add to cart
Holds:
Bottle (5 L)
Color:
Yellow
Price (SGD)
489.00
EA
Add to cart
The new modular Bottle Carrying System fits standard laboratory bottles that have a GL 45 neck finish and is compatible with four bottle sizes: 2, 5, 10 and 20 liters. It is ergonomically designed, and offers health and safety benefits to the user and allows easy and safe transport of bottles around the laboratory or production facility. It makes pouring liquids from the larger bottles more comfortable and safer. All materials are fully autoclavable. The collar is only removable by cutting – so attachment is 100 % secure in use.
  • The ergonomical design offers health and safety benefits and allows easy and safe transport of bottles
  • It makes pouring liquids from larger bottles more comfortable and safer
  • The collar is only removable by cutting what makes the attachment 100 % secure in use
Specifications
AutoclavableAutoclavable
ColorYellow
HoldsBottle (5 L)
MaterialSilicone Rubber, Polypropylene, Synthetic Rubber
For Use With (Application)Safe transport of laboratory bottles; Decanting of liquids
Quantity1 Ea.
SizeGL 45
Diameter (Metric) Outer194 mm
Unit SizeEA