Micropipettes, BLAUBRAND™, intraMark, DE-M, CE-IVD
Actual product may vary
Micropipettes, BLAUBRAND™, intraMark, DE-M, CE-IVD
Micropipettes, BLAUBRAND™, intraMark, DE-M, CE-IVD
Fisherbrand
Micropipettes, BLAUBRAND™, intraMark, DE-M, CE-IVD
Constructed of disposable glass and conforms to DIN ISO 7550 standards. BRAND™ Blaubrand™ IntraMARK™ Disposable Micropipets include ring mark, and are CE marked according to guideline EU IVDR 2017/746. All surfaces are fire polished for smoothness and to minimize pipet adapter wear. Color-coded to ISO for clear identification.
Have Questions?
Catalog NumberColorAccuracyGraduations
FKS15744037White≤±0.30%1, 2, 3, 4, 5 μL
Catalog number FKS15744037
Price (SGD)
-
Color:
White
Accuracy:
≤±0.30%
Graduations:
1, 2, 3, 4, 5 μL
  • DIN ISO 7550. 
  • DE-M marking for volumes greater than 5 μl
  • Calibrated to contain (TC, In) 
  • Color code to ISO for clear identification
  • CE marked according to guideline EU IVDR 2017/746
Specifications
Accuracy≤±0.30%
CalibrationTo Contain
Certifications/ComplianceDIN ISO 7550
MaterialGlass
Graduations1, 2, 3, 4, 5 μL
Quantity250/Box
ColorWhite
Length (Metric)125 mm