Diamond Antistatic Weighing Boats
Diamond Antistatic Weighing Boats
Actual product may vary
Diamond Antistatic Weighing Boats
Provide accurate pour-outs with minimal sample loss. Diamond Antistatic Weighing Boats are flexible polystyrene boats featuring smooth surfaces.
Have Questions?
Catalog NumberColor
FKS15718197White
Catalog number FKS15718197
Price (SGD)
90.00
PK
Add to cart
Color:
White
Price (SGD)
90.00
PK
Add to cart
  • Diamond-shaped boats that flex for accurate pouring.
  • Wide, flat bottom to resist tipping
  • 2 colors
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • Antistatic.
  • Temperature range -10°C to +70°C (14°F to 158°F)
  • Can be used with microwave moisture analyzers
  • All are FDA/food-grade compliant
  • Pack of 500
  • Disposable
Specifications
AutoclavableNot Autoclavable
Bottom ShapeFlat
ColorWhite
DisposableSingle-use
MaterialPolystyrene
ShapeDiamond
SterilityNon-sterile
TypeWeighing Boat
For Use With (Equipment)Microwave Moisture Analyzer
Temperature Range (English)14°F to 158°F
Temperature Range (Metric)-10°C to +70°C
Length (English)2.8 in.
Length (Metric)71 mm
Width (English)1.8 in.
Width (Metric)46 mm
Height (Metric)14 mm
Height (English)0.6 in.
Unit SizePK