Upright ULT Freezer Drawer Racks for Centrifuge Tubes
Actual product may vary
Fisherbrand
Upright ULT Freezer Drawer Racks for Centrifuge Tubes
Fisherbrand inventory control racks are ideal for all cold storage application. Provide an ideal alternative for storing large quantities of samples in 15mL or 50mL centrifuges tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
FKS15709559Conical Tubes (78 x 50 mL)
FKS15799549Conical Tubes (160 x 15 mL)
Catalog number FKS15709559
Price (SGD)
508.00
Each
Holds:
Conical Tubes (78 x 50 mL)
Price (SGD)
508.00
Each
  • For 15mL or 50mL centrifuges tubes
  • 2 drawers, holds between 78 and 160 tubes
  • Made of high quality stainless steel
Specifications
HoldsConical Tubes (78 x 50 mL)
MaterialStainless Steel
For Use With (Application)50ml conical tubes
For Use With (Equipment)Upright ULT Freezer
Dimensions (L x W x H)559 x 252 x 140 mm
Product TypeDrawer Rack
Unit SizeEach