Stainless Steel Forceps
Actual product may vary
Fisherbrand
Stainless Steel Forceps
Straight, blunt round ends. Nickel-Electro™ Stainless Steel Forceps contain serrated jaws.
Have Questions?
Catalog NumberLength (Metric)
FKS15699310300 mm
Catalog number FKS15699310
Price (SGD)
40.00
Each
Length (Metric):
300 mm
Price (SGD)
40.00
Each
  • Stainless Steel Forceps
  • Straight, serrated jaws
  • Blunt round ends
  • 300mm Length
Specifications
Product TypeLaboratory forceps/tweezers
Length (English)12 in.
Length (Metric)300 mm
MaterialStainless Steel
Dimensions (L x W)300 mm
No. per Pack1 Ea.
Tip StyleBlunt
Unit SizeEach