SlimTube™ Cryogenic Vials
Actual product may vary
Fisherbrand
SlimTube™ Cryogenic Vials
Optimizes the capacity with 23.5% compared to conventional storage solutions. Tenak™ SlimTube™ Cryogenic Vials are the first of their kind with external thread to be stored in standard 10 x 10 cryoboxes.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)
FKS156550195.0 mL88.5 mm
Catalog number FKS15655019
Price (SGD)
-
Capacity (Metric):
5.0 mL
Height (Metric):
88.5 mm
  • Unique cryotube is the first of its kind with external thread
  • The external thread limits sample contamination and is the preferred choice when selecting a cryotube
  • Cap is designed with single-handed quick seal closure for easy handling.The steep thread design allows tightening and removal of the cap with only half a turn
  • Constructed of specially formulated polypropylene to withstand temperatures to –196°C
  • The polypropylene used for both cap and tube has the same coefficient of expansion, ensuring an equally secure seal both at room temperature and at low cryogenic temperatures
  • Cap includes a conical lip seal which makes it self-sealing,100% leak proof, and user-friendly
  • Two indicators for maximum recommended filling. One indicator for storing at -80°C, and one for storing in liquid nitrogen in the vapour phase at -196°C
  • Star foot design allows the tube to be self-standing.
  • Compatible with common working stations on the market, and suitable for singlehanded manipulation
Specifications
Diameter (Metric)12 mm
Capacity (Metric)5.0 mL
Bottom ShapeStarfoot
MaterialSpecially Formulated Polypropylene
Thread StyleExternal
IncludesWhite label for marking
No. per Pack500
Packaging5 resealable bags of 100
RNase FreeYes
Temperature Range (Metric)As low as -196°C
SterilitySterile
GraduatedYes
Dnase FreeYes
Height (Metric)88.5 mm
Unit SizePack