Use as a sampling spoon or to measure or transfer small amounts. Buerkle™ SteriPlast™ Spoon Spatulas can also be used as a scraper or for mixing and stirring.
Have Questions?
Catalog Number
Capacity (Metric)
FKS15614114
0.5 mL
Catalog number FKS15614114
Price (SGD)
284.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
0.5 mL
Price (SGD)
284.00
PK
Add to cart
The spoon spatula is a double spatula with a 0.5 mL spoon and a 17 mm spatula blade, total length 178 mm
PS, specially designed for disposable use, resistant to chemicals and corrosion-proof
Production, assembly and packaging according to clean room class 7
Complies with EU food requirements and FDA regulations
Optionally packaged individually and sterilised by gamma rays
Specifications
MaterialPolystyrene
Capacity (Metric)0.5 mL
DisposableSingle-use
SterilitySterile
Width (Metric)17 mm spoon blade
Unit SizePK
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
0 results displayed, search above for a specific certificate