Vortex-Genie™ 2
Vortex-Genie™ 2
Actual product may vary
Fisherbrand
Vortex-Genie™ 2
Mixes gently, vigorously, and anywhere in between. Scientific Industries SI™ Vortex-Genie™ 2 speeds range from 600 to 3200rpm.
Have Questions?
Catalog NumberModelPlug Type
FKS15557335SI-0266UK Plug
Catalog number FKS15557335
Price (SGD)
1,916.00
EA
Add to cart
Model:
SI-0266
Plug Type:
UK Plug
Price (SGD)
1,916.00
EA
Add to cart
  • Variable speed for precise mixing
  • Broad range of attachments available for most tubes, plates and other containers
  • Hands-Free or Touch On control
  • May be used in cold rooms and incubators
Specifications
Speed Range600 to 3200 rpm
Electrical Requirements230 V, 60 Hz
Voltage230 V
Product TypeVortex Mixer
Plug TypeUK Plug
Frequency60 Hz
Depth (Metric)165 mm
Height (Metric)165 mm
Width (Metric)122 mm
Weight (Metric)4 kg
ModelSI-0266
IncludesPop-off cup, 76mm platform
Operating ModesHands-free, Touch on
Unit SizeEA