Check Order Status
Desiccator with Glass Knob
Actual product may vary
Fisherbrand
Desiccator with Glass Knob
Made from Simax 3.3 borosilicate glass. Fisherbrand Desiccator with Glass Knob is designed for variety of vacuum work.
Have Questions?
Catalog Number
Product Type
Diameter (Metric) Inner
Diameter (Metric) Outer
Height (Metric)
FKS15534645
Desiccator
285 mm
380 mm
438 mm
Catalog number
FKS15534645
Price (SGD)
1,710.00
EA
Add to cart
Product Type:
Desiccator
Diameter (Metric) Inner:
285 mm
Diameter (Metric) Outer:
380 mm
Height (Metric):
438 mm
Price (SGD)
1,710.00
EA
Add to cart
Made from SIMAX 3.3 borosilicate glass
Designed for variety of vacuum work
Used for desiccation of samples, storage of materials sensitive to humidity
Specifications
Certifications/Compliance
ISO 13 130, DIN 12 491
Material
Simax Borosilicate Glass 3.3
Product Type
Desiccator
Includes
Glass knob
Height (Metric)
438 mm
Diameter (Metric) Inner
285 mm
Diameter (Metric) Outer
380 mm