Search Fisher Scientific
| Catalog Number | Thread Size (Metric) |
|---|---|
| FKS15499644 | GLS 80 |
Bottle mixing system for use with overhead laboratory mixers, Materials used: PP and PTFE. Flexible modular system with a central ground joint 29/32 fitting. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be used by changing the inserts. Sterile pressure equalisation is possible by using the syringe filter. Unused ports can be closed with a blind cap.