Borosilicate Glass Narrow Neck Erlenmeyer Flasks
Borosilicate Glass Narrow Neck Erlenmeyer Flasks
Actual product may vary
Borosilicate Glass Narrow Neck Erlenmeyer Flasks
Constructed of borosilicate glass. Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks are conical and graduated.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1549909350 mL
FKS15409103100 mL
FKS154691033000 mL
FKS15419103200 mL
FKS15429103250 mL
FKS15439103500 mL
FKS154491031000 mL
Catalog number FKS15499093
Price (SGD)
97.00
PK
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
97.00
PK
Add to cart
Erlenmeyer, graduated.
Specifications
Certifications/ComplianceISO 1773
Diameter (Metric) Outer Neck22 mm
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Height (Metric)90 mm
ShapeConical
GraduatedYes
Mouth StyleNarrow
Diameter (Metric) Outer51 mm
Unit SizePK