Straight Containers
Actual product may vary
Fisherbrand
Straight Containers
Designed for sample collection, storage and transportation to laboratories. Gosselin™ Straight Containers feature a leakproof cap.
Have Questions?
Catalog NumberCapacity (Metric)SterilityClosure Color
FKS1547881460 mLNon-sterileWhite
Catalog number FKS15478814
Price (SGD)
388.00
Case
Capacity (Metric):
60 mL
Sterility:
Non-sterile
Closure Color:
White
Price (SGD)
388.00
Case
  • Shatter resistant polypropylene is specially designed for use in the food, dairy, and brewing industries
  • LDPE screw caps included
  • Leakproof inner lip prevents drops between threads
  • Sterility Assurance Level based on ISO 11137 : STERILE SAL 10-3
Specifications
Product TypeSpecimen Container
Closure IncludedYes
MaterialPolypropylene
SterilityNon-sterile
Capacity (Metric)60 mL
Certifications/ComplianceISO 11137
Closure ColorWhite
Closure MaterialLDPE
Closure TypeScrew Cap
ColorNatural
Diameter (Metric)33 mm
For Use With (Application)Bacteriological applications
Height (Metric)70 mm
IncludesLDPE screw cap with leakproof inner lip
LabelNo Label
No. per Pack700/Pk.
Unit SizeCase