Feature heavy wall construction for increased mechanical strength. Fisherbrand™ Borosilicate Glass Squat Form Heavy Duty Beakers are made of full 3.3 expansion borosilicate glass and feature an 'easy pour' spout.
Have Questions?
Catalog Number
Capacity (Metric)
FKS15476123
250 mL
Catalog number FKS15476123
Price (SGD)
-
Capacity (Metric):
250 mL
Heavy wall construction for increased mechanical strength