Check Order Status
HDPE Wide Neck Square Reagent Bottles
Actual product may vary
Fisherbrand
HDPE Wide Neck Square Reagent Bottles
The Wide-neck reagent bottle is particularly suited for storing, packing, taking samples, sampling and transport of granules and paste-like media.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS15475574
50 mL
FKS15425574
100 mL
FKS15376418
250 mL
FKS11817702
500 mL
FKS15316438
1000 mL
5 Options
Catalog number
FKS15475574
Price (SGD)
11.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
11.00
EA
Add to cart
HDPE, transparent
PP tamper-evident screw cap, blue
UN approval for bulk goods
Specifications
Capacity (Metric)
50 mL
Color
Transparent
Diameter (Metric) Outer Neck
28 mm
Dimensions (L x W x H)
36 x 36 x 89 mm
Diameter (Metric) Inner Neck
21 mm
For Use With (Application)
Foodstuffs
Material
HDPE
Neck ID (Metric)
21 mm
Weight (Metric)
0.018 kg
Width (Metric)
36 mm
Closure Color
Blue
Closure Included
Yes
Height (Metric)
89 mm
Closure Material
Polypropylene
Closure Type
Screw Cap